Gene Transcripts

Gene Symbol
Mixl1
Entrez Gene ID
27217
Gene Name
Mix1 homeobox-like 1 (Xenopus laevis)
Gene Aliases
Mm1, Mml
Chromosome Location
Chr. 1: 180693043 - 180709055 on Build GRCm38
UniGene ID
Mm.103647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013729.3 NP_038757.1 2 1178
GenBank mRNA AF154573.1 - 2 1178
AF201959.1 - 2 1128
AK138043.1 - 1 494

View Full Product Details