Gene Transcripts

Gene Symbol
Timm8a1
Entrez Gene ID
30058
Gene Name
translocase of inner mitochondrial membrane 8A1
Gene Aliases
DXHXS1274E, Ddp1, Fci-12, Tim8a, Timm8a
Chromosome Location
Chr. X: 134537258 - 134541629 on Build GRCm38
UniGene ID
Mm.214504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013898.2 NP_038926.1 2 229
GenBank mRNA AB031055.1 - 2 229
AF150081.1 - 2 220
AK011402.1 - 2 264
AK088903.1 - 2 222
AK090079.1 - 2 268
AK165573.1 - 2 224
BC004796.1 - 2 217 3588797
BC094631.1 - 2 238 3419065

View Full Product Details