Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013867.2 | NP_038895.1 | 4 | 771 | |
GenBank mRNA | AF179566.1 | - | 4 | 719 | |
AK143894.1 | - | 4 | 559 | ||
AK158241.1 | - | 4 | 773 | ||
AK167578.1 | - | 4 | 590 | ||
BC023930.1 | - | 4 | 760 | 5324365 |