Gene Transcripts

Gene Symbol
Abcf2
Entrez Gene ID
27407
Gene Name
ATP-binding cassette, sub-family F (GCN20), member 2
Gene Aliases
0710005O05Rik, Drr3, E430001O06
Chromosome Location
Chr. 5: 24565341 - 24577467 on Build GRCm38
UniGene ID
Mm.21629
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190443.1 NP_001177372.1 5, 6 988
NM_013853.2 NP_038881.1 5, 6 991
GenBank mRNA AK087990.1 - 5, 6 893
AK137233.1 - 5, 6 923
AK147937.1 - 5, 6 946
AK151545.1 - 5, 6 947
BC003300.1 - 4, 5 883 3583837

View Full Product Details