Gene Transcripts

Gene Symbol
Zpbp
Entrez Gene ID
53604
Gene Name
zona pellucida binding protein
Gene Aliases
4930486K01Rik, Iam38, Sp38
Chromosome Location
Chr. 11: 11279624 - 11463630 on Build GRCm38
UniGene ID
Mm.9500
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185153.1 NP_001172082.1 6 793
NM_015785.2 NP_056600.1 6 964
GenBank mRNA AK006274.1 - 3 306
AK018889.1 - 3 308
AK018921.1 - 3 308
AK029541.1 - 3 308
BC139450.1 - 6 933 8862472
BC139453.1 - 6 933 8862475
BC145628.1 - 6 762 9054196
D17569.1 - 6 953

View Full Product Details