Gene Transcripts

Gene Symbol
Hist1h1e
Entrez Gene ID
50709
Gene Name
histone cluster 1, H1e
Gene Aliases
H1-4, H1.4, H1e, H1f4, H1s-4, H1var2
Chromosome Location
Chr. 13: 23621777 - 23622558 on Build GRCm38
UniGene ID
Mm.170587
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015787.4 NP_056602.1 1 600
GenBank mRNA BC089600.1 - 600

View Full Product Details