Gene Transcripts
- Gene Symbol
- Tpmt
- Entrez Gene ID
- 22017
- Gene Name
- thiopurine methyltransferase
- Gene Aliases
- AW106912
- Chromosome Location
- Chr. 13: 47021987 - 47043338 on Build GRCm38
- UniGene ID
- Mm.10169
- Species
- Mus musculus
- Gene Symbol
- Gm8479
- Entrez Gene ID
- 667141
- Gene Name
- thiopurine S-methyltransferase pseudogene
- Gene Aliases
- EG667141
- Chromosome Location
- Chr. 6: 65328120 - 65329149 on Build GRCm38
- UniGene ID
- Mm.425331
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF037046.1 |
- |
1 |
490 |
|
AF037047.1 |
- |
1 |
485 |
|
View Full Product Details