Gene Transcripts

Gene Symbol
Svs2
Entrez Gene ID
53878
Gene Name
seminal vesicle secretory protein 2
Gene Aliases
BB115391, Semg1, SvsII
Chromosome Location
Chr. 2: 164235929 - 164238341 on Build GRCm38
UniGene ID
Mm.99395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017390.4 NP_059086.3 2 217
GenBank mRNA AK035204.1 - 2 142
AK040724.1 - 2 143
AK090151.1 - 2 139
AK090302.1 - 2 143
AK136679.1 - 2 141
AK144285.1 - 2 142
AK144389.1 - 2 142
AK144429.1 - 2 142
AK144524.1 - 2 139
AK165704.1 - 2 142
AK165713.1 - 2 142
AK165727.1 - 2 139
AK165753.1 - 2 142
AK165825.1 - 2 142
BB871464.1 - 2 209
BC107275.1 - 2 142 40052650
BC107276.1 - 2 142
DV056729.1 - 2 251

View Full Product Details