Gene Transcripts

Gene Symbol
Sult2b1
Entrez Gene ID
54200
Gene Name
sulfotransferase family, cytosolic, 2B, member 1
Gene Aliases
AI326997, BB173635, ST2B1, SULT2B
Chromosome Location
Chr. 7: 45721174 - 45759555 on Build GRCm38
UniGene ID
Mm.271634
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017465.2 NP_059493.2 7 1113
GenBank mRNA AF026072.1 - 7 1080
BC009811.1 - 7 1155 4008307
BC009813.1 - 7 1043 4039050

View Full Product Details