Gene Transcripts

Gene Symbol
NAT8
Entrez Gene ID
9027
Gene Name
N-acetyltransferase 8 (putative)
Gene Aliases
ATase2, CCNAT, CML1, GLA, Hcml1, TSC501, TSC510
Chromosome Location
Chr. 2: 73640723 - 73642410 on Build GRCh38
UniGene ID
Hs.14637
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003960.3 NP_003951.3 2 844
GenBank mRNA AB013094.1 - 2 845
AB019551.1 - 2 803
AF187813.1 - 2 854

View Full Product Details