Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018817.2 | NP_061287.2 | 4 | 1128 | |
GenBank mRNA | AF088884.1 | - | 4 | 1108 | |
AK035854.1 | - | 4 | 1105 | ||
AK042332.1 | - | 4 | 1107 | ||
AK077878.1 | - | 4 | 1106 | ||
AK083488.1 | - | 4 | 1106 | ||
AK146767.1 | - | 4 | 1122 | ||
AK154020.1 | - | 4 | 1111 | ||
AK161570.1 | - | 4 | 1048 | ||
AK169465.1 | - | 4 | 1119 | ||
BC029078.1 | - | 4 | 1063 | 3992325 |