Gene Transcripts

Gene Symbol
Impa1
Entrez Gene ID
55980
Gene Name
inositol (myo)-1(or 4)-monophosphatase 1
Gene Aliases
2610002K09Rik, 2900059K10Rik, AI325909
Chromosome Location
Chr. 3: 10313540 - 10331439 on Build GRCm38
UniGene ID
Mm.183042
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018864.5 NP_061352.2 6 530
GenBank mRNA AF042730.1 - 6 475
AK011770.1 - 6 468
AK078406.1 - 6 479
AK083238.1 - 6 464
AK145472.1 - 467
AK147778.1 - 6 490
AK152774.1 - 6 481
AK153457.1 - 6 497
AK160203.1 - 6 468
AK160467.1 - 6 462
AK165977.1 - 6 472
AK168457.1 - 6 505
BC049080.1 - 6 471 6419835
BC055722.1 - 5 425 6413389

View Full Product Details