Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164186.1 | NP_001157658.1 | 2 | 872 | |
NM_019446.4 | NP_062319.1 | 1 | 1155 | ||
GenBank mRNA | AB043980.1 | - | 1 | 1170 | |
AF264026.1 | - | 3 | 930 | ||
AJ237590.1 | - | 1 | 430 | ||
AK036312.1 | - | 1 | 607 | ||
BC055731.1 | - | 2 | 873 | 6413504 |