Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001038700.2 | NP_001033789.1 | 3 | 394 | |
NM_001177648.1 | NP_001171119.1 | 3 | 394 | ||
NM_001177649.1 | NP_001171120.1 | 3 | 394 | ||
NM_019406.3 | NP_062279.1 | 3 | 394 | ||
GenBank mRNA | AK122308.1 | - | 3 | 394 | |
AK157135.1 | - | 3 | 385 | ||
AK170497.1 | - | 3 | 344 | ||
AK172100.1 | - | 3 | 338 | ||
BC003867.1 | - | 3 | 325 | 3490113 |