Gene Transcripts

Gene Symbol
Pdlim4
Entrez Gene ID
30794
Gene Name
PDZ and LIM domain 4
Gene Aliases
Ril
Chromosome Location
Chr. 11: 54054928 - 54069032 on Build GRCm38
UniGene ID
Mm.21830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019417.3 NP_062290.2 5 660
GenBank mRNA AB221672.1 - 5 575
BC030068.1 - 5 589 5144166
Y08361.1 - 5 611

View Full Product Details