Gene Transcripts

Gene Symbol
KCNJ13
Entrez Gene ID
3769
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
KIR1.4, KIR7.1, LCA16, SVD
Chromosome Location
Chr. 2: 232765802 - 232776565 on Build GRCh38
UniGene ID
Hs.467338
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172416.1 NP_001165887.1 3 2363
NM_001172417.1 NP_001165888.1 3 2366
NM_002242.4 NP_002233.2 3 2599
GenBank mRNA AJ007557.1 - 3 2578
BC037290.2 - 3 2599 4817527

View Full Product Details