Gene Transcripts

Gene Symbol
Ramp3
Entrez Gene ID
56089
Gene Name
receptor (calcitonin) activity modifying protein 3
Gene Aliases
AI850306
Chromosome Location
Chr. 11: 6658533 - 6677476 on Build GRCm38
UniGene ID
Mm.39884
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019511.3 NP_062384.1 3 906
GenBank mRNA AF146524.1 - 3 907
AF209907.1 - 3 866
AJ250491.1 - 3 907
AK005513.1 - 4 1022

View Full Product Details