Gene Transcripts

Gene Symbol
Tmem45a
Entrez Gene ID
56277
Gene Name
transmembrane protein 45a
Gene Aliases
C630002M10Rik, M32486, p19.5
Chromosome Location
Chr. 16: 56805161 - 56886458 on Build GRCm38
UniGene ID
Mm.262328
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019631.3 NP_062605.2 6 2399
GenBank mRNA AK049837.1 - 6 2400

View Full Product Details