Gene Transcripts

Gene Symbol
Styx
Entrez Gene ID
56291
Gene Name
serine/threonine/tyrosine interaction protein
Gene Aliases
0610039A20Rik, hStyxb
Chromosome Location
Chr. 14: 45351186 - 45376884 on Build GRCm38
UniGene ID
Mm.202561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019637.3 NP_062611.2 11 1015
GenBank mRNA AK002822.1 - 957
AK030122.1 - 11 1015
AK031186.1 - 1 77
AK042741.1 - 12 1070
AK154136.1 - 11 1110
AK166043.1 - 11 998
BC038608.1 - 11 965 5012303
BC096674.1 - 10 925 5144003
U34973.1 - 12 1119

View Full Product Details