Gene Transcripts
- Gene Symbol
- Clptm1
- Entrez Gene ID
- 56457
- Gene Name
- cleft lip and palate associated transmembrane protein 1
- Gene Aliases
- HS9, N14
- Chromosome Location
- Chr. 7: 19631580 - 19665030 on Build GRCm38
- UniGene ID
- Mm.290960
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019649.2 |
NP_062623.2
|
2, 3 |
218 |
|
GenBank mRNA |
AK148080.1 |
- |
2, 3 |
200 |
|
AK152410.1 |
- |
2, 3 |
208 |
|
AK152809.1 |
- |
2, 3 |
208 |
|
AK165742.1 |
- |
2, 3 |
218 |
|
AK171901.1 |
- |
2, 3 |
209 |
|
AK194191.1 |
- |
2, 3 |
143 |
|
AK207441.1 |
- |
3, 4 |
232 |
|
AK218417.1 |
- |
3, 4 |
341 |
|
BC022172.1 |
- |
2, 3 |
209 |
4193279 |
BY288640.1 |
- |
2, 3 |
222 |
|
D67067.1 |
- |
2, 3 |
205 |
|
View Full Product Details