Gene Transcripts
- Gene Symbol
- Aldh18a1
- Entrez Gene ID
- 56454
- Gene Name
- aldehyde dehydrogenase 18 family, member A1
- Gene Aliases
- 2810433K04Rik, AI429789, Pycs
- Chromosome Location
- Chr. 19: 40550257 - 40588463 on Build GRCm38
- UniGene ID
- Mm.233117
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF056573.1 |
- |
5, 6 |
608 |
|
AF056574.1 |
- |
5, 6 |
608 |
|
View Full Product Details