Gene Transcripts
- Gene Symbol
- Anapc7
- Entrez Gene ID
- 56317
- Gene Name
- anaphase promoting complex subunit 7
- Gene Aliases
- APC7, AW545589
- Chromosome Location
- Chr. 5: 122422444 - 122444912 on Build GRCm38
- UniGene ID
- Mm.37341
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019805.4 |
NP_062779.3
|
6, 7 |
959 |
|
GenBank mRNA |
AF076607.2 |
- |
6, 7 |
923 |
|
AK030775.1 |
- |
6, 7 |
924 |
|
AK049968.1 |
- |
6, 7 |
898 |
|
AK137286.1 |
- |
6, 7 |
923 |
|
AK150764.1 |
- |
6, 7 |
923 |
|
AK150891.1 |
- |
6, 7 |
923 |
|
AK153114.1 |
- |
6, 7 |
923 |
|
AK159523.1 |
- |
6, 7 |
891 |
|
AK164202.1 |
- |
6, 7 |
928 |
|
BC006635.1 |
- |
6, 7 |
912 |
3482261 |
View Full Product Details