Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019819.3 | NP_062793.2 | 3 | 690 | |
GenBank mRNA | AF120113.1 | - | 1 | 416 | |
AK009744.1 | - | 3 | 703 | ||
AK079362.1 | - | 3 | 691 | ||
AK163738.1 | - | 3 | 735 | ||
AK166230.1 | - | 3 | 688 | ||
AK166840.1 | - | 3 | 686 | ||
BC002130.1 | - | 3 | 655 | 3592420 | |
BC052836.1 | - | 3 | 710 | 30049007 |