Gene Transcripts

Gene Symbol
Slc35g3
Entrez Gene ID
56293
Gene Name
solute carrier family 35, member G3
Gene Aliases
Amac1
Chromosome Location
Chr. 11: 69759884 - 69761844 on Build GRCm38
UniGene ID
Mm.6211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019871.2 NP_063924.2 2 787
GenBank mRNA AF016712.1 - 2 637
AK077181.1 - 2 913
BC137893.1 - 2 667 8860912
BC137894.1 - 2 667 8860913

View Full Product Details