Gene Transcripts

Gene Symbol
STK10
Entrez Gene ID
6793
Gene Name
serine/threonine kinase 10
Gene Aliases
LOK, PRO2729
Chromosome Location
Chr. 5: 172042070 - 172188342 on Build GRCh38
UniGene ID
Hs.744005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005990.3 NP_005981.3 1, 2 455
GenBank mRNA AB015718.1 - 1, 2 205
AK313350.1 - 2, 3 210
BC070077.1 - 1, 2 369 5267449
BI459912.1 - 1, 2 374
DA507768.1 - 1, 2 455

View Full Product Details