Gene Transcripts

Gene Symbol
Acp6
Entrez Gene ID
66659
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
5730559A09Rik, ACPL1, AU022842, mPACPL1
Chromosome Location
Chr. 3: 97158752 - 97176576 on Build GRCm38
UniGene ID
Mm.101368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019800.4 NP_062774.2 6 1065
GenBank mRNA AB030038.1 - 6 1058
AF216223.1 - 6 542
AK002783.1 - 6 979
AK077729.1 - 6 1053
AK160271.1 - 6 984
AK165356.1 - 6 1066
AK171757.1 - 6 980
BC047276.1 - 6 805 6487551

View Full Product Details