Gene Transcripts
- Gene Symbol
- Mpp6
- Entrez Gene ID
- 56524
- Gene Name
- membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6)
- Gene Aliases
- P55t, Pals2
- Chromosome Location
- Chr. 6: 50110241 - 50198598 on Build GRCm38
- UniGene ID
- Mm.248361
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164733.1 |
NP_001158205.1
|
3, 4 |
407 |
|
NM_001164734.1 |
NP_001158206.1
|
3, 4 |
417 |
|
NM_019939.2 |
NP_064323.2
|
3, 4 |
417 |
|
GenBank mRNA |
AF161181.1 |
- |
3, 4 |
281 |
|
AF199009.1 |
- |
3, 4 |
413 |
|
AF199010.1 |
- |
3, 4 |
413 |
|
AK144434.1 |
- |
3, 4 |
418 |
|
AK164549.1 |
- |
3, 4 |
408 |
|
BC042868.2 |
- |
3, 4 |
364 |
|
BC046421.1 |
- |
3, 4 |
364 |
|
BC069908.1 |
- |
3, 4 |
406 |
|
BC106140.1 |
- |
3, 4 |
364 |
|
BC138666.1 |
- |
3, 4 |
275 |
8861687 |
BC145363.1 |
- |
3, 4 |
275 |
9053921 |
BC145364.1 |
- |
3, 4 |
265 |
9053922 |
View Full Product Details