Gene Transcripts

Gene Symbol
Dnajb8
Entrez Gene ID
56691
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 8
Gene Aliases
1700016F14Rik, 1700031F10Rik, AI326313, mDj6
Chromosome Location
Chr. 6: 88222268 - 88223256 on Build GRCm38
UniGene ID
Mm.272871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019964.1 NP_064348.1 1 368
GenBank mRNA AB028856.1 - 1 368
AK006026.1 - 1 352
BC049591.1 - 1 346 6773959
BC061112.1 - 1 351 6772873

View Full Product Details