Gene Transcripts

Gene Symbol
Habp4
Entrez Gene ID
56541
Gene Name
hyaluronic acid binding protein 4
Gene Aliases
4933413D03Rik, 4933428J01Rik
Chromosome Location
Chr. 13: 64161866 - 64186537 on Build GRCm38
UniGene ID
Mm.40989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019986.3 NP_064370.2 8 1988
GenBank mRNA AF227684.1 - 8 1924
AK144206.1 - 8 1967
AK146525.1 - 8 1961
BC082806.1 - 7 4284

View Full Product Details