Gene Transcripts

Gene Symbol
Slco1b2
Entrez Gene ID
28253
Gene Name
solute carrier organic anion transporter family, member 1b2
Gene Aliases
7330442B20Rik, OATP-C, OATP2, Oatp1b2, Oatp4, Slc21a10, Slc21a6, lst-1, mlst-1
Chromosome Location
Chr. 6: 141629452 - 141686646 on Build GRCm38
UniGene ID
Mm.272223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020495.1 NP_065241.1 4 322
GenBank mRNA AB031959.1 - 4 322

View Full Product Details