Gene Transcripts

Gene Symbol
Arhgap31
Entrez Gene ID
12549
Gene Name
Rho GTPase activating protein 31
Gene Aliases
5830477L08Rik, AU041750, Cdgap, D330026I07Rik
Chromosome Location
Chr. 16: 38598343 - 38713035 on Build GRCm38
UniGene ID
Mm.370848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020260.2 NP_064656.2 9, 10 1430
GenBank mRNA AF151363.1 - 9, 10 1071
AK138417.1 - 9, 10 1670
AK149975.1 - 9, 10 1266
AK150109.1 - 9, 10 1266
AK155729.1 - 9, 10 1250
AK170685.1 - 9, 10 1250
BC138870.1 - 9, 10 1235 8861892

Gene Symbol
A130004G07Rik
Entrez Gene ID
320687
Gene Name
RIKEN cDNA A130004G07 gene
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK037298.1 - 1 336

View Full Product Details