Gene Transcripts

Gene Symbol
Ap4m1
Entrez Gene ID
11781
Gene Name
adaptor-related protein complex AP-4, mu 1
Gene Aliases
4930443L05Rik, Ap4m4
Chromosome Location
Chr. 5: 138172021 - 138178687 on Build GRCm38
UniGene ID
Mm.113205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021392.4 NP_067367.3 4 475
GenBank mRNA AF242858.1 - 4 343
AK031638.1 - 4 478
AK165187.1 - 3 711
BC011174.1 - 4 360 4207033

View Full Product Details