Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025571.2 | NP_001020742.1 | 5 | 882 | |
NM_021309.3 | NP_067284.1 | 5 | 882 | ||
GenBank mRNA | AF203343.1 | - | 5 | 889 | |
AK037489.1 | - | 5 | 604 | ||
AK172182.1 | - | 5 | 634 | ||
BC034847.1 | - | 5 | 614 | 3327113 | |
U69490.1 | - | 5 | 592 |