Gene Transcripts

Gene Symbol
Rbak
Entrez Gene ID
57782
Gene Name
RB-associated KRAB zinc finger
Gene Aliases
Chromosome Location
Chr. 5: 143172182 - 143180842 on Build GRCm38
UniGene ID
Mm.320419
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045482.2 NP_001038947.1 5 2830
NM_021326.3 NP_067301.1 5 2930
GenBank mRNA AK051009.1 - 5 2931
BC059913.1 - 3 2689

View Full Product Details