Gene Transcripts

Gene Symbol
SYNPR
Entrez Gene ID
132204
Gene Name
synaptoporin
Gene Aliases
SPO
Chromosome Location
Chr. 3: 63200604 - 63616924 on Build GRCh38
UniGene ID
Hs.648668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130003.1 NP_001123475.1 5 811
NM_144642.4 NP_653243.1 4 824
XM_017005731.1 XP_016861220.1 5 577
XM_017005732.1 XP_016861221.1 5 811
GenBank mRNA AF411860.2 - 4 573
AK122831.1 - 5 601
AK293628.1 - 4 523
AK309017.1 - 4 606
AK312465.1 - 4 729
AL834457.1 - 4 553
BC022518.1 - 4 573 4792167
CR936762.1 - 5 732

View Full Product Details