Gene Transcripts

Gene Symbol
Drg2
Entrez Gene ID
13495
Gene Name
developmentally regulated GTP binding protein 2
Gene Aliases
AI255295, AI461605
Chromosome Location
Chr. 11: 60454617 - 60468724 on Build GRCm38
UniGene ID
Mm.41803
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021354.1 NP_067329.1 13 1426
GenBank mRNA AJ243590.1 - 13 1426
BC082564.1 - 13 1382 30551418

View Full Product Details