Gene Transcripts

Gene Symbol
Thap11
Entrez Gene ID
59016
Gene Name
THAP domain containing 11
Gene Aliases
2810036E22Rik, AB041579, CTG-B43a, CTG-B45d, Ronin
Chromosome Location
Chr. 8: 105855103 - 105856950 on Build GRCm38
UniGene ID
Mm.286598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021513.2 NP_067488.1 1 1067
GenBank mRNA AB041579.1 - 1 1028
AK165981.1 - 1 1028
AK170457.1 - 1 1039
BC003949.1 - 1 620
BC138966.1 - 1 979 8861988
BC138987.1 - 1 979 8862009

View Full Product Details