Gene Transcripts

Gene Symbol
Dexi
Entrez Gene ID
58239
Gene Name
dexamethasone-induced transcript
Gene Aliases
1810029J14Rik, AI836170, AW413143, D16Bwg0586e, Myle
Chromosome Location
Chr. 16: 10530207 - 10543122 on Build GRCm38
UniGene ID
Mm.353006
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021428.4 NP_067403.2 2 1359
GenBank mRNA AF152470.2 - 2 1060
AK007644.1 - 2 1360
BC017551.1 - 2 1346 4910742

View Full Product Details