Gene Transcripts

Gene Symbol
Tnni1
Entrez Gene ID
21952
Gene Name
troponin I, skeletal, slow 1
Gene Aliases
2700018B22Rik, AI747285, ssTnI
Chromosome Location
Chr. 1: 135783035 - 135810989 on Build GRCm38
UniGene ID
Mm.44379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112702.1 NP_001106173.1 9 588
NM_021467.5 NP_067442.1 9 636
GenBank mRNA AA230582.1 - 9 636
AA271187.1 - 9 587
AJ242874.1 - 9 636
AK012258.1 - 9 619
BC023170.1 - 8 700 4949804

View Full Product Details