Gene Transcripts

Gene Symbol
Sqrdl
Entrez Gene ID
59010
Gene Name
sulfide quinone reductase-like (yeast)
Gene Aliases
0610039J17Rik, 4930557M22Rik, SQOR
Chromosome Location
Chr. 2: 122764928 - 122809569 on Build GRCm38
UniGene ID
Mm.28986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162503.1 NP_001155975.1 5 584
NM_021507.5 NP_067482.4 5 903
GenBank mRNA AF174535.1 - 5 903
AK008290.1 - 5 550
AK016166.1 - 5 536
AK075598.1 - 5 585
AK149948.1 - 5 555
BC011153.1 - 4 522 4196280
BC028247.1 - 5 572

View Full Product Details