Gene Transcripts
- Gene Symbol
- Smu1
- Entrez Gene ID
- 74255
- Gene Name
- smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans)
- Gene Aliases
- 2600001O03Rik, 2610203K23Rik, AB044414, AI845086, AW556129, Bwd
- Chromosome Location
- Chr. 4: 40735649 - 40757885 on Build GRCm38
- UniGene ID
- Mm.289929
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021535.4 |
NP_067510.3
|
2, 3 |
328 |
|
GenBank mRNA |
AB044414.1 |
- |
2, 3 |
257 |
|
AK003493.1 |
- |
2, 3 |
315 |
|
AK011140.1 |
- |
2, 3 |
327 |
|
AK019174.1 |
- |
2, 3 |
287 |
|
AK145982.1 |
- |
2, 3 |
327 |
|
AK159635.1 |
- |
2, 3 |
315 |
|
AK168813.1 |
- |
2, 3 |
322 |
|
BC020034.1 |
- |
2, 3 |
270 |
|
BC057446.1 |
- |
2, 3 |
289 |
3155760 |
BY036103.1 |
- |
2, 3 |
330 |
|
View Full Product Details