Gene Transcripts

Gene Symbol
Gpr88
Entrez Gene ID
64378
Gene Name
G-protein coupled receptor 88
Gene Aliases
AW061286, Strg
Chromosome Location
Chr. 3: 116249654 - 116253498 on Build GRCm38
UniGene ID
Mm.40025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022427.2 NP_071872.1 2 2799
GenBank mRNA AB042408.1 - 2 2769
BC051941.1 - 1 2415 5708059

View Full Product Details