Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023219.2 | NP_075708.2 | 12 | 1425 | |
GenBank mRNA | AF251268.1 | - | 12 | 1428 | |
AF411960.2 | - | 12 | 1345 | ||
AK008310.1 | - | 2 | 125 | ||
AK008368.1 | - | 2 | 125 | ||
AK008500.1 | - | 2 | 124 | ||
AK144296.1 | - | 1368 | |||
BC109133.1 | - | 12 | 1364 | ||
BC109134.1 | - | 12 | 1364 | ||
BC127612.1 | - | 12 | 1364 | ||
BC127615.1 | - | 12 | 1364 | 40061657 | |
BC128465.1 | - | 12 | 1364 |