Gene Transcripts
- Gene Symbol
- Sdhb
- Entrez Gene ID
- 67680
- Gene Name
- succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
- Gene Aliases
- 0710008N11Rik
- Chromosome Location
- Chr. 4: 140961271 - 140979197 on Build GRCm38
- UniGene ID
- Mm.246965
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023374.3 |
NP_075863.2
|
4, 5 |
559 |
|
GenBank mRNA |
AF095936.1 |
- |
1, 2 |
110 |
|
AK003052.1 |
- |
4, 5 |
453 |
|
AK003533.1 |
- |
4, 5 |
443 |
|
AK009660.1 |
- |
4, 5 |
453 |
|
AK153045.1 |
- |
4, 5 |
436 |
|
AK169252.1 |
- |
4, 5 |
450 |
|
AK169839.1 |
- |
4, 5 |
452 |
|
AK190493.1 |
- |
1, 2 |
31 |
|
AK196072.1 |
- |
2, 3 |
216 |
|
BC013509.1 |
- |
4, 5 |
454 |
4225025 |
BC018580.1 |
- |
4, 5 |
418 |
|
BC051934.1 |
- |
4, 5 |
559 |
5688898 |
DQ403000.1 |
- |
3, 4 |
350 |
|
View Full Product Details