Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023627.1 | NP_076116.1 | 3, 4 | 362 | |
GenBank mRNA | AF288525.1 | - | 3, 4 | 362 | |
AK005029.2 | - | 3, 4 | 340 | ||
AK079323.1 | - | 3, 4 | 339 | ||
AK137089.1 | - | 3, 4 | 355 | ||
BC003458.1 | - | 3, 4 | 353 | 2648362 |