Gene Transcripts

Gene Symbol
Tollip
Entrez Gene ID
54473
Gene Name
toll interacting protein
Gene Aliases
4930403G24Rik, 4931428G15Rik
Chromosome Location
Chr. 7: 141881582 - 141902465 on Build GRCm38
UniGene ID
Mm.103551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023764.3 NP_076253.1 4 592
GenBank mRNA AJ242971.1 - 4 596
AK015062.1 - 535
AK016482.1 - 4 593
AK045422.1 - 4 576
AK049263.1 - 4 614
AK049394.1 - 4 538
AK078391.1 - 602
AK161312.1 - 4 579
AK189903.1 - 3 366
BC053515.1 - 4 579
BC062139.1 - 4 573 6334946

View Full Product Details