Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301367.1 | NP_001288296.1 | 3 | 398 | |
NM_024255.3 | NP_077217.2 | 2 | 300 | ||
GenBank mRNA | AJ293845.1 | - | 3 | 321 | |
AK011910.1 | - | 2 | 258 | ||
AK085899.1 | - | 2 | 269 | ||
AK136773.1 | - | 2 | 267 | ||
AK144948.1 | - | 2 | 216 | ||
AK145230.1 | - | 2 | 160 | ||
AK202946.1 | - | 1 | 15 | ||
AK205169.1 | - | 1 | 15 | ||
AK211849.1 | - | 1 | 15 | ||
AK215396.1 | - | 1 | 15 | ||
AY725196.1 | - | 2 | 157 | ||
BC002211.1 | - | 2 | 173 | 3490303 | |
BY327952.1 | - | 2 | 301 |