Gene Transcripts

Gene Symbol
Retnlb
Entrez Gene ID
57263
Gene Name
resistin like beta
Gene Aliases
9030012B21Rik, Fizz2, RELMbeta, Relmb, Xcp3
Chromosome Location
Chr. 16: 48816856 - 48818892 on Build GRCm38
UniGene ID
Mm.21123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023881.4 NP_076370.3 2 357
GenBank mRNA AF290871.1 - 2 359
AF323083.1 - 2 244
AK020240.1 - 2 245
BC022650.1 - 2 227 4216606
DQ157777.1 - 1 116

View Full Product Details