Gene Transcripts

Gene Symbol
Riok3
Entrez Gene ID
66878
Gene Name
RIO kinase 3
Gene Aliases
1200013N13Rik, D18Ertd331e, E130306C24Rik, Sudd
Chromosome Location
Chr. 18: 12128850 - 12157367 on Build GRCm38
UniGene ID
Mm.28551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024182.4 NP_077144.2 5 528
GenBank mRNA AK004748.1 - 5 620
AK131635.1 - 5 532
AK134327.1 - 5 620
AK147074.1 - 5 529
AK167309.1 - 5 624
AK170882.1 - 5 608
BC002255.1 - 2 58
BC033271.1 - 5 566 4016197

View Full Product Details