Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024499.1 | NP_078775.1 | 4, 5 | 366 | |
GenBank mRNA | AK039966.1 | - | 3, 4 | 361 | |
AK051005.1 | - | 4, 5 | 365 | ||
AK079168.1 | - | 4, 5 | 376 | ||
AK165557.1 | - | 4, 5 | 390 | ||
AK189623.1 | - | 1, 2 | 75 | ||
AK193443.1 | - | 1, 2 | 77 | ||
BC003836.1 | - | 4, 5 | 366 | 3486148 |